Chloromethyltrimethylsilane, 98+%
Chloromethyltrimethylsilane, 98+%
Chloromethyltrimethylsilane, 98+%
Thermo Scientific Chemicals

Chloromethyltrimethylsilane, 98+%

CAS: 2344-80-1 | C4H11ClSi | 122.67 g/mol
Quantity:
25 g
100 g
Catalog number 194761000
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS2344-80-1
IUPAC Name(chloromethyl)trimethylsilane
Molecular FormulaC4H11ClSi
InChI KeyOOCUOKHIVGWCTJ-UHFFFAOYSA-N
SMILESC[Si](C)(C)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4165 to 1.4185 (20°C, 589 nm)
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only