2-Acetyl-1-naphthol, 99%
2-Acetyl-1-naphthol, 99%
2-Acetyl-1-naphthol, 99%
Thermo Scientific Chemicals

2-Acetyl-1-naphthol, 99%

CAS: 711-79-5 | C12H10O2 | 186.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
View all
Plastic bottle
Catalog number 195000250
Price (TWD)
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS711-79-5
IUPAC Name1-(1-hydroxynaphthalen-2-yl)ethan-1-one
Molecular FormulaC12H10O2
InChI KeyJBGJVMVWYWUVOW-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C2C=CC=CC2=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Form)Crystalline powder
Melting point97°C to 100°C
Infrared spectrumConforms
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only