Forma™ Inner Door Kits
Forma™ Inner Door Kits
Actual product may vary
Thermo Scientific™
Forma™ Inner Door Kits
Reduce cold air loss and improve temperature recovery after door closings when you choose factory-installed Thermo Scientific™ Forma™ Inner Door Kits for your ultra-low upright freezer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
1950548Forma 7000 and 900 Series, 23.0 cu. ft. Uprights
1950546Forma 7000 and 900 Series, 13.0 cu. ft. Uprights
1950547Forma 7000 and 900 Series, 17.3 cu. ft. Uprights
195240Forma -40°C Upright Lab Freezers
1950529Forma 7000 and 900 Series, 28.0 cu. ft. Uprights
Catalog number 1950548
Price (MXN)
-
For Use With (Equipment):
Forma 7000 and 900 Series, 23.0 cu. ft. Uprights
Reduce cold air loss and improve temperature recovery after door closings with Thermo Scientific™ Forma™ Inner Door Kits, a factory-installed door alternative option for your ultra-low upright freezer.
Four interior doors are already included on our -86°C uprights. The five inner door option for 8600 and 900 Series includes five powder-coated inner doors and one additional shelf.
Ordering Information:
Factory installed. Must be ordered with the refrigerator or freezer.
Specifications
Exterior FinishPowder-coated Steel
For Use With (Equipment)Forma 7000 and 900 Series, 23.0 cu. ft. Uprights
TypeUltra-Low Freezer Inner-Door
Capacity (English)23 cu. ft.
Capacity (Metric)651.2 L
No. of Doors5
Shelves1
StyleUpright
Temperature Range-86°C
Unit SizeEach