Thermo Scientific Chemicals

Palladium on barium sulfate, 5% Pd, unreduced, Thermo Scientific Chemicals

Catalog number: 195060500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Palladium on barium sulfate, 5% Pd, unreduced, Thermo Scientific Chemicals

Catalog number: 195060500
50 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
7440-05-3
IUPAC Name
palladium
Molecular Formula
Pd
InChI Key
KDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES
[Pd]
Palladium (Pd)
4.50 to 5.50 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text