Thermo Scientific Chemicals

Sodium tetrachloropalladate(II), approx. 36% Pd, Thermo Scientific Chemicals

Catalog number: 195130050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Sodium tetrachloropalladate(II), approx. 36% Pd, Thermo Scientific Chemicals

Catalog number: 195130050
5 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
13820-53-6
IUPAC Name
disodium tetrachloropalladiumbis(ylium)
Molecular Formula
Cl4Na2Pd
InChI Key
ABKQFSYGIHQQLS-UHFFFAOYSA-J
SMILES
[Na+].[Na+].Cl[Pd++](Cl)(Cl)Cl
Silver (Ag)
=<100 ppm
Copper (Cu)
=<50 ppm
Appearance (Form)
Powder or granules and/or chunks
Total trace metal impurities
=<300 ppm (Pt + Ru + Rh + Ir)
Iron (Fe)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text