Palladium(II) bromide, 39.8-40.1% Pd
Palladium(II) bromide, 39.8-40.1% Pd
Palladium(II) bromide, 39.8-40.1% Pd
Thermo Scientific Chemicals

Palladium(II) bromide, 39.8-40.1% Pd

CAS: 13444-94-5 | Br2Pd | 266.23 g/mol
数量:
2 g
10 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 195190020
価格(JPY)
-
数量:
2 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13444-94-5
IUPAC Namepalladium(2+) dibromide
Molecular FormulaBr2Pd
InChI KeyINIOZDBICVTGEO-UHFFFAOYSA-L
SMILES[Br-].[Br-].[Pd++]
さらに表示
Appearance (Color)Dark red-brown to black
Silver (Ag)=<20 ppm
Gold (Au)=<20 ppm
Iron (Fe)=<20 ppm
Ruthenium (Ru)=<100 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only