POROS™ CaptureSelect™ IgM Affinity Matrix

Catalog number: 195289005

 Related applications: Chromatography

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

POROS™ CaptureSelect™ IgM Affinity Matrix is intended for the purification of human IgM, with no binding to other immunoglobulins.

Features of this affinity matrix include:
• IgM purification from plasma and serum
• Purification of monoclonal IgM from cell cultures
• Efficient purification of IgM with high yields and retained activity
• Excellent scalability
• Non-animal derived
• High flow rates
• Cross-reactivity towards mouse and rat IgM

POROS CaptureSelect IgM Affinity Matrix binds to the Fc part of IgM. The high selectivity and yields obtained using IgM Affinity Matrix enable a robust, fast, and efficient purification process with excellent purity obtained in one step.

Free of animal components
Thermo Fisher Scientific POROS CaptureSelect products contain affinity ligands created by a proprietary technology based on camelid-derived single-domain antibody fragments. The ligand is a 13-kDa single-domain fragment that comprises the 3 CDRs that form the antigen binding domain and is efficiently produced by the yeast Saccharomyces cerevisiae in a production process free of animal components.

Main characteristics:

Matrix: Cross-linked poly(styrene-divinylbenzene)
Average particle size: 50 µm
Ligand: CaptureSelect IgM affinity ligand
Ligand coupling method: Carbonyldiimidazole (CDI)
Binding capacity: ~6 g/L resin (determined with polyclonal human IgM from serum)
Elution conditions: Acidic elution with 0.1 M glycine, pH 3.0
Mechanical resistance: 100 bar (1,450 psi, 10 MPa)
Formulation buffer: 20% (v/v) ethanol
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Column Type: Proprietary Resin
Immobilized Phase: Anti-IgM
Product Line: POROS™ CaptureSelect™
Product Size: 5 mL

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols