Potassium hexachloroplatinate(IV), ca. 40% Pt
Potassium hexachloroplatinate(IV), ca. 40% Pt
Potassium hexachloroplatinate(IV), ca. 40% Pt
Thermo Scientific Chemicals

Potassium hexachloroplatinate(IV), ca. 40% Pt

CAS: 16921-30-5 | Cl6K2Pt | 485.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 195350010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16921-30-5
IUPAC Namedipotassium hexachloroplatinumdiuide
Molecular FormulaCl6K2Pt
InChI KeyDPAIVKJGTXERIM-UHFFFAOYSA-H
SMILES[K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder and/or chunks
Appearance (Color)Yellow to light orange
Platinum (Pt)39.4 to 40.6 % (Pt)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only