Potassium tetrachloroplatinate(II), 46-47% Pt
Potassium tetrachloroplatinate(II), 46-47% Pt
Potassium tetrachloroplatinate(II), 46-47% Pt
Thermo Scientific Chemicals

Potassium tetrachloroplatinate(II), 46-47% Pt

CAS: 10025-99-7 | Cl4H4K2Pt | 419.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 195360050
Price (USD)
584.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
584.00
Each
Chemical Identifiers
CAS10025-99-7
IUPAC Namedipotassium tetrachloroplatinumdiuide
Molecular FormulaCl4H4K2Pt
InChI KeyJCAXKWJYKUEOFO-UHFFFAOYSA-N
SMILESCl.Cl.Cl.Cl.[K+].[K+].[Pt--]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Total trace metal impurities=<500 ppm (referring to Pt)
Gravimetry46 to 47 % (Pt)
Appearance (Color)Red-brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only