Evans Blue
Evans Blue
Evans Blue
Thermo Scientific Chemicals

Evans Blue

CAS: 314-13-6 | C34H24N6Na4O14S4 | 960.79 g/mol
Have Questions?
Catalog NumberQuantityPackaging
19555025025 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 195550250
Price (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS314-13-6
IUPAC Nametetrasodium (6E)-4-amino-6-[2-(4'-{2-[(2E)-8-amino-1-oxo-5,7-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-5-oxo-5,6-dihydronaphthalene-1,3-disulfonate
Molecular FormulaC34H24N6Na4O14S4
InChI KeyKBNIFDASRCWYGC-GXNXWABVSA-J
SMILES[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1/C=CC2=C(C=C(C(N)=C2C1=O)S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2/C=CC3=C(C=C(C(N)=C3C2=O)S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Loss on drying=<10 %
UV Lambda max313 to 317 nm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only