Search
| 產品號碼 | Type |
|---|---|
| 195893 | Sample Probe, Factory Installed |
| 195892 | Sample Probe, Customer Installed |
Measure sample temperature or top chamber temperature in upright freezers with Thermo Scientific™ Forma™ Sample Probe Kits. Use probe kit with Forma 8600 Series Upright Freezers.