Cobalt standard solution, for AAS, 1 mg/ml Co in 2 to 5% HNO3
Cobalt standard solution, for AAS, 1 mg/ml Co in 2 to 5% HNO3
Cobalt standard solution, for AAS, 1 mg/ml Co in 2 to 5% HNO3
Thermo Scientific Chemicals

Cobalt standard solution, for AAS, 1 mg/ml Co in 2 to 5% HNO3

CAS: 10141-05-6 | CoN2O6 | 182.941 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 mL
500 mL
產品號碼 195921000
價格 (TWD)
-
Quantity:
100 mL
Request bulk or custom format
規格
CAS10141-05-6
CAS7732-18-5
ChEBICHEBI:86209
Chemical Name or MaterialCobalt standard solution
Formula Weight58.93
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopy
RUO – Research Use Only