Thermo Scientific™

Hafnium standard solution, for AAS, 1 mg/ml Hf in 5% HF, Thermo Scientific™

100mL, Plastic bottle, Each
Thermo Scientific™

Hafnium standard solution, for AAS, 1 mg/ml Hf in 5% HF, Thermo Scientific™

100mL, Plastic bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
7440-58-6, 7664-39-3, 7732-18-5
Appearance (Form)
Solution
Strength
990 to 1010 ug/ml of Hf(4+) in 5% HF
Appearance (Color)
Clear colorless

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
For atomic adsorption spectroscopy
RUO – Research Use Only

Figures