Thermo Scientific™

Lithium standard solution, for AAS, 1 mg/ml Li in 2% HCl, Thermo Scientific™

500mL, Plastic bottle, Each
Thermo Scientific™

Lithium standard solution, for AAS, 1 mg/ml Li in 2% HCl, Thermo Scientific™

500mL, Plastic bottle, Each
Quantity
Packaging
Catalog number: 196085000
Price (USD)
137.60 / Each
Your Price:
Quantity
-

Chemical Identifiers

CAS
7439-93-2, 7647-01-0, 7732-18-5
Appearance (Form)
Clear solution
Strength
995 to 1005 ug/ml of Li (1+) in 2% HCl (20°C)

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
For atomic adsorption spectroscopy
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures