MabCaptureC Affinity Matrix
Thermo Scientific™

MabCaptureC Affinity Matrix

Thermo Scientific™ MabCaptureC Affinity Matrix is an agarose-based, high-binding-capacity Protein A chromatography resin with a rProtein A ligand produced in yeast.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
196366205L5 L
1963662250250 mL
196366201L1 L
196366210L10 L
Número de catálogo 196366205L
Precio (MXN)
-
Cantidad:
5 L

MabCaptureC Affinity Matrix is an agarose-based, high-binding-capacity Protein A chromatography resin with a rProtein A ligand produced in yeast. The engineered rProtein A binds at the CH2-CH3 interface of the Fc-region of IgGs and shows cross binding to the VH3 domain of IgGs.

MabCaptureC Affinity Matrix is suitable for the purification of monoclonal antibodies.The rProtein A ligand is immobilized on a highly cross-linked and uniform-sized agarose-based bead with an average particle size of 75 μm. The uniform particle size delivers superior performance characteristics compared to traditional Protein A resins.

Features of the MabCaptureC Affinity Matrix include:

  • High dynamic binding capacity for mAbs of >50 g/L at 4.8 min residence time
  • Purification of mAbs and a subpopulation of scFv's and Fab fragments containing VH3 binding domains
  • Clean and sanitize with standard sodium hydroxide agents for chromatography products
  • Excellent scalability
  • Non-animal derived

Main characteristics

  • Matrix: agarose-based, epoxide activated
  • Average particle size: 60–80 μm
  • Ligand: recombinant C domain of Protein A
  • Ligand coupling method: epoxide coupling
  • Binding capacity: >50 g IgG/L resin at 4.8 min residence time and 10% breakthrough
  • Elution conditions: 0.1 M glycine pH 3.0, or 20–100 mM sodium citrate or acetate, pH 3.0
  • Cleaning conditions: 0.2 M sodium hydroxide for 15 minutes every cycle
  • Operating pressure: ≤3 bar
  • Pressure flow: >500 cm/h at 3 bar in a 2.6 x 20 cm column
  • Formulation buffer: 20% (v/v) ethanol

For process development and resin screening, MabCaptureC™ is also available in the following formats:

  • MabCaptureC™ MiniChrom Column, 1 mL (Cat. No. 5943662001)
  • MabCaptureC™ MiniChrom Column for process development, 5 mL (5943662005)
  • MabCaptureC™ RoboColumn, 200 μL (5943662200)
  • MabCaptureC™ RoboColumn, 600 μL (5943662600)

Para su uso en investigación o procesos de fabricación posteriores. No apto para uso diagnóstico ni para la administración directa en seres humanos ni en animales.

Especificaciones
Capacidad de unión>50 g IgG/L
Para utilizar con (aplicación)Monoclonal antibody purification
FormulaciónSuspension in 20% ethanol
Tipo de ligandoEngineered rProtein A
MatrizAgarose
Presión máx. de funcionamiento≤3 bar
Cantidad5 L
ObjetivoCH2-CH3 interface Fc-region
Capacidad (métrico)>50 g IgG/L
FormularioResin Slurry
Tamaño de partículas70 μm
TipoAffinity Matrix
Unit SizeEach
Contenido y almacenamiento
• Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light