MabCaptureC Affinity Matrix
Thermo Scientific™

MabCaptureC Affinity Matrix

Thermo Scientific™ MabCaptureC Affinity Matrix is an agarose-based, high-binding-capacity Protein A chromatography resin with a rProtein A ligand produced in yeast.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
196366210L10 L
1963662250250 mL
196366201L1 L
196366205L5 L
Catalog number 196366210L
Price (MXN)
-
Quantity:
10 L

MabCaptureC Affinity Matrix is an agarose-based, high-binding-capacity Protein A chromatography resin with a rProtein A ligand produced in yeast. The engineered rProtein A binds at the CH2-CH3 interface of the Fc-region of IgGs and shows cross binding to the VH3 domain of IgGs.

MabCaptureC Affinity Matrix is suitable for the purification of monoclonal antibodies.The rProtein A ligand is immobilized on a highly cross-linked and uniform-sized agarose-based bead with an average particle size of 75 μm. The uniform particle size delivers superior performance characteristics compared to traditional Protein A resins.

Features of the MabCaptureC Affinity Matrix include:

  • High dynamic binding capacity for mAbs of >50 g/L at 4.8 min residence time
  • Purification of mAbs and a subpopulation of scFv's and Fab fragments containing VH3 binding domains
  • Clean and sanitize with standard sodium hydroxide agents for chromatography products
  • Excellent scalability
  • Non-animal derived

Main characteristics

  • Matrix: agarose-based, epoxide activated
  • Average particle size: 60–80 μm
  • Ligand: recombinant C domain of Protein A
  • Ligand coupling method: epoxide coupling
  • Binding capacity: >50 g IgG/L resin at 4.8 min residence time and 10% breakthrough
  • Elution conditions: 0.1 M glycine pH 3.0, or 20–100 mM sodium citrate or acetate, pH 3.0
  • Cleaning conditions: 0.2 M sodium hydroxide for 15 minutes every cycle
  • Operating pressure: ≤3 bar
  • Pressure flow: >500 cm/h at 3 bar in a 2.6 x 20 cm column
  • Formulation buffer: 20% (v/v) ethanol

For process development and resin screening, MabCaptureC™ is also available in the following formats:

  • MabCaptureC™ MiniChrom Column, 1 mL (Cat. No. 5943662001)
  • MabCaptureC™ MiniChrom Column for process development, 5 mL (5943662005)
  • MabCaptureC™ RoboColumn, 200 μL (5943662200)
  • MabCaptureC™ RoboColumn, 600 μL (5943662600)
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
For Use With (Application)Monoclonal antibody purification
FormatContainer
Ligand TypeEngineered rProtein A
MatrixAgarose
Max. Operating Pressure≤3 bar
Quantity10 L
Shipping ConditionWet Ice
TargetCH2-CH3 interface Fc-region
Binding Capacity>50 g IgG/L
FormBulk Media
Particle Size70 μm
Product LineMabCaptureC
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.