Iodine, 99.5%, extra pure, resublimed
Iodine, 99.5%, extra pure, resublimed
Iodine, 99.5%, extra pure, resublimed
Acros Organics

Iodine, 99.5%, extra pure, resublimed

CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number 196560025
also known as 19656
HSN 28012000
Price (INR)
118,264.00
Each
Quantity:
2.5 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
118,264.00
Each
Chemical Identifiers
CAS7553-56-2
IUPAC Namediiodine
Molecular FormulaI2
InChI KeyPNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILESII
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matterPasses test
Appearance (Color)Dark grey
Appearance (Form)Metallic spheres
Titration Iodometric>=99.4 %
Chloride and Bromide=<0.005 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only