Cuivre, 99 %, poudre, max. 106μ m (maille de -140), Thermo Scientific Chemicals
Cuivre, 99 %, poudre, max. 106μ m (maille de -140), Thermo Scientific Chemicals
Cuivre, 99 %, poudre, max. 106μ m (maille de -140), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cuivre, 99 %, poudre, max. 106μ m (maille de -140), Thermo Scientific Chemicals

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
25 g
500 g
2.5 kg
Conditionnement:
Flacon en verre
Référence 196570250
Prix (EUR)
26,20
Each
Quantité:
25 g
Conditionnement:
Flacon en verre
Grand volume ou format personnalisé
Prix (EUR)
26,20
Each
Identifiants chimiques
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
Voir plus
Appearance (Color)Red to red-brown
Appearance (Form)Powder
Assay>=98.5 % (Cu)
Average particle size10 to 28 µm
Particle size=<106 µm
Voir plus
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only