Potassium cyanide, 98+%, for analysis, Thermo Scientific Chemicals
Potassium cyanide, 98+%, for analysis, Thermo Scientific Chemicals
Potassium cyanide, 98+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium cyanide, 98+%, for analysis, Thermo Scientific Chemicals

数量:
100 g
250 g
500 g
2.5 kg
製品番号(カタログ番号) 196600025
価格(JPY)
-
数量:
2.5 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS151-50-8
IUPAC Namepotassium iminomethanide
Molecular FormulaCKN
InChI KeyNNFCIKHAZHQZJG-UHFFFAOYSA-N
SMILES[K+].[C-]#N
さらに表示
Appearance (Form)Crystalline or granular powder
Water=<0.1 %
Free alkali (KOH)0.2 to 0.6 %
Titration Argentometric98.0 to 102.0 %
Carbonate (CO3)0.2 to 0.7 % (as K2CO3)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only