Ammonium carbonate, for analysis
Ammonium carbonate, for analysis
Ammonium carbonate, for analysis
Thermo Scientific Chemicals

Ammonium carbonate, for analysis

CAS: 506-87-6 | CH8N2O3 | 96.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 196651000
Price (USD)
53.70
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
53.70
Each
Chemical Identifiers
CAS506-87-6
IUPAC Namecarbonic acid diamine
Molecular FormulaCH8N2O3
InChI KeyPRKQVKDSMLBJBJ-UHFFFAOYSA-N
SMILESN.N.OC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or chunks
Sulfate (SO4)=<50 ppm
Iron (Fe)=<10 ppm
Heavy metals (as Pb)=<10 ppm
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only