Copper(II) carbonate basic, pure, 54-56% Cu
Copper(II) carbonate basic, pure, 54-56% Cu
Copper(II) carbonate basic, pure, 54-56% Cu
Copper(II) carbonate basic, pure, 54-56% Cu
Thermo Scientific Chemicals

Copper(II) carbonate basic, pure, 54-56% Cu

CAS: 12069-69-1 | CCuO3·CuO2H2 | 221.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 196675000
Price (USD)
76.80
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
76.80
Each
Chemical Identifiers
CAS12069-69-1
IUPAC Namedicopper(2+) dihydroxide carbonate
Molecular FormulaCH2Cu2O5
InChI KeyZMMDPCMYTCRWFF-UHFFFAOYSA-J
SMILES[OH-].[OH-].[Cu++].[Cu++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay54 to 56 % (Cu)
Appearance (Color)Green
Appearance (Form)Crystalline powder

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only