Europium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Europium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Europium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Thermo Scientific Chemicals

Europium(III) nitrate hexahydrate, 99.9%, (trace metal basis)

CAS: 10031-53-5 | EuN3O9·6H2O | 446.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1966800505 gGlass bottle
Catalog number 196680050
Price (USD)
149.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
149.00
Each
Chemical Identifiers
CAS10031-53-5
IUPAC Nameeuropium(3+) hexahydrate trinitrate
Molecular FormulaEuH12N3O15
InChI KeyJVYYYCWKSSSCEI-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Eu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total conc. of imp.as REO=<0.1 %
Appearance (Form)Crystalline solid
Appearance (Color)Semitransparent
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only