Fluoroboric acid, 50 wt% solution in water
Fluoroboric acid, 50 wt% solution in water
Fluoroboric acid, 50 wt% solution in water
Thermo Scientific Chemicals

Fluoroboric acid, 50 wt% solution in water

CAS: 16872-11-0 | BF4H | 87.812 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 196700250
Price (USD)
70.20
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
70.20
Each
Chemical Identifiers
CAS16872-11-0
IUPAC Namehydrogen tetrafluoroboranuide
Molecular FormulaBF4H
InChI KeyODGCEQLVLXJUCC-UHFFFAOYSA-O
SMILES[H+].F[B-](F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Solution
Iron (Fe)=<30 ppm
Heavy metals (as Pb)=<50 ppm
AppearanceMay develop some turbidity or precipitate
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only