Hidracina hidrato, 100 % (hidracina, 64 %), Thermo Scientific Chemicals
Hidracina hidrato, 100 % (hidracina, 64 %), Thermo Scientific Chemicals
Hidracina hidrato, 100 % (hidracina, 64 %), Thermo Scientific Chemicals
Hidracina hidrato, 100 % (hidracina, 64 %), Thermo Scientific Chemicals
Hidracina hidrato, 100 % (hidracina, 64 %), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hidracina hidrato, 100 % (hidracina, 64 %), Thermo Scientific Chemicals

CAS: 10217-52-4 | H4N2 | 32.05 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
100 g
500 g
Envase:
Frasco de vidrio
Número de catálogo 196711000
Precio (USD)
-
Cantidad:
100 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS10217-52-4
IUPAC Namehydrazine
Molecular FormulaH4N2
InChI KeyOAKJQQAXSVQMHS-UHFFFAOYSA-N
SMILESNN
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Clear colorless
Titration with HCl63.6 to 64.6 % (N2H4)
Density(20°C) 1.030 to 1.033 g/ml
Appearance (Form)Liquid
Refractive index1.4285 to 1.4315 (20°C, 589 nm)
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reactivo de reducción
RUO – Research Use Only