Sodium thiocyanate, 98%, extra pure
Sodium thiocyanate, 98%, extra pure
Sodium thiocyanate, 98%, extra pure
Thermo Scientific Chemicals

Sodium thiocyanate, 98%, extra pure

CAS: 540-72-7 | CNNaS | 81.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 196765000
Price (USD)
92.65
Online exclusive
103.00
Save 10.35
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
92.65
Online exclusive
103.00
Save 10.35
Each
Chemical Identifiers
CAS540-72-7
IUPAC Namesodium cyanosulfanide
Molecular FormulaCNNaS
InChI KeyVGTPCRGMBIAPIM-UHFFFAOYSA-M
SMILES[Na+].[S-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=97.5 % (on dried substance)
Iron (Fe)=<2 ppm
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals and/or chunks
Loss on drying=<2.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only