Sodium hexafluorophosphate, 98.5+%, pure
Sodium hexafluorophosphate, 98.5+%, pure
Sodium hexafluorophosphate, 98.5+%, pure
Thermo Scientific Chemicals

Sodium hexafluorophosphate, 98.5+%, pure

Sodium hexafluorophosphate, 98.5+%, pure, Quantity: 50g, Packaging: Plastic bottle, CAS: 21324-39-0, White to Gray, Grade: Pure, Melting Point: 200.0 deg.C, Molecular Formula: F6NaP, Molecular Weight: 167.95, Percent Purity: 98.5+% | CAS: 21324-39-0 | F6NaP | 167.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Plastic bottle
Catalog number 196822500
Price (USD)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS21324-39-0
IUPAC Namesodium hexafluoro-λ⁵-phosphanuide
Molecular FormulaF6NaP
InChI KeyKMADQUOCJBLXRP-UHFFFAOYSA-N
SMILES[Na+].F[P-](F)(F)(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light grey
Appearance (Form)Crystalline powder
Assay>=98.50 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only