Cloruro de estaño(II), 98 %, anhidro, Thermo Scientific Chemicals
Cloruro de estaño(II), 98 %, anhidro, Thermo Scientific Chemicals
Cloruro de estaño(II), 98 %, anhidro, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cloruro de estaño(II), 98 %, anhidro, Thermo Scientific Chemicals

CAS: 7772-99-8 | Cl2Sn | 189.61 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
2.5 kg
Envase:
Frasco de vidrio
Número de catálogo 196985000
Precio (USD)
-
Cantidad:
500 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS7772-99-8
IUPAC Nameλ²-tin(2+) dichloride
Molecular FormulaCl2Sn
InChI KeyAXZWODMDQAVCJE-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Sn++]
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White to off-white
Iron (Fe)=<50 ppm
Appearance (Form)Crystalline powder or flakes
Lead (Pb)=<75 ppm
Titration Iodimetric97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only