Vanadium(III) chloride, 97%
Vanadium(III) chloride, 97%
Vanadium(III) chloride, 97%
Thermo Scientific Chemicals

Vanadium(III) chloride, 97%

Vanadium(III) chloride, 97%, Cl3V, CAS Number-7718-98-1, Vanadium trichloride, 100g, 96% min. (Argentometry), Black to Blue, 3.0000g/mL, 231-744-6, 13,289, 157.3, GHS Signal Word: Danger | CAS: 7718-98-1 | Cl3V | 157.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 197000250
Price (USD)
169.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
169.00
Each
Chemical Identifiers
CAS7718-98-1
IUPAC Namevanadium(3+) trichloride
Molecular FormulaCl3V
InChI KeyHQYCOEXWFMFWLR-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[V+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=96.0 %
Appearance (Form)Powder
Insoluble matter=<0.5 % (in water)
Appearance (Color)Dark blue to violet to black
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only