Molecular sieves 3A, 4 to 8 mesh
Molecular sieves 3A, 4 to 8 mesh
Molecular sieves 3A, 4 to 8 mesh
Thermo Scientific Chemicals

Molecular sieves 3A, 4 to 8 mesh

CAS: 308080-99-1 | AlNaO6Si2 | 202.135 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 kg
Packaging:
View all
Glass bottle
Catalog number 197245000
Price (USD)
95.60
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
95.60
Each
Chemical Identifiers
CAS308080-99-1
IUPAC NameMolecular sieves 3A
MDL NumberMFCD00131613
SpecificationsSpecification SheetSpecification Sheet
Water adsorption>=21 % (50%rH, 25°C, 1bar)
Particle size>=95 % (3-5 mm)
Appearance (Color)Light grey to beige
Appearance (Form)Beads
Bulk density680 to 800 g/l
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent
RUO – Research Use Only