Molecular sieves 4A, 8 to 12 mesh
Molecular sieves 4A, 8 to 12 mesh
Molecular sieves 4A, 8 to 12 mesh
Thermo Scientific Chemicals

Molecular sieves 4A, 8 to 12 mesh

CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 kg
25 kg
Packaging:
View all
Fibre Drum
Catalog number 197270250
Price (EUR)
-
Quantity:
25 kg
Packaging:
Fibre Drum
Request bulk or custom format
Chemical Identifiers
CAS70955-01-0
IUPAC NameMolecular sieves
Synonym2-[2-acetamido-3-(3-fluorophenyl)propanamido]-5-methoxy-4-(methoxycarbonyl)-3-methyl-5-oxopentanoic acid
MDL NumberMFCD00131613
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light grey to beige
Appearance (Form)Beads
Bulk density680 to 800 g/l
Residual water=<1.5 % (550°C, 2h)
Water adsorption>=21 % (50%rH, 25°C, 1bar)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent
RUO – Research Use Only