Thermo Scientific™

Molecular sieves 13X, 8 to 12 mesh, Thermo Scientific™

500g, Glass bottle, Each
Thermo Scientific™

Molecular sieves 13X, 8 to 12 mesh, Thermo Scientific™

500g, Glass bottle, Each
Quantity
Packaging
Catalog number: 197315000
Price (USD)
Price: 73.00 / Each
Your Price:
Quantity
-

Chemical Identifiers

CAS
63231-69-6
Appearance (Color)
Beige to brown
Bulk density
>=600 g/l
Particle size
8 to 12 mesh
Appearance (Form)
Beads
Residual water
=<1.5 % (550°C, 2 h)

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Drying agent
RUO – Research Use Only

Figures