Ruthenium(IV) oxide hydrate, > 54% Ru
Ruthenium(IV) oxide hydrate, > 54% Ru
Ruthenium(IV) oxide hydrate, > 54% Ru
Ruthenium(IV) oxide hydrate, > 54% Ru
Ruthenium(IV) oxide hydrate, > 54% Ru
Thermo Scientific Chemicals

Ruthenium(IV) oxide hydrate, > 54% Ru

CAS: 32740-79-7 | O2Ru | 133.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 197430050
Price (EUR)
337,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
337,00
Each
Chemical Identifiers
CAS32740-79-7
IUPAC Namedioxoruthenium
Molecular FormulaO2Ru
InChI KeyWOCIAKWEIIZHES-UHFFFAOYSA-N
SMILESO=[Ru]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=54 % (Ru)
Appearance (Color)Black
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only