Thermo Scientific Chemicals

Osmium tetroxide, 2.5 wt.% solution in tert-Butanol, stabilized, Thermo Scientific Chemicals

Catalog number: 197450250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

Osmium tetroxide, 2.5 wt.% solution in tert-Butanol, stabilized, Thermo Scientific Chemicals

Catalog number: 197450250
25 mL, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
20816-12-0, 75-65-0
Appearance (Color)
Colorless to yellow
Appearance (Form)
Fused mass or solution
Strength
>=2.5 wt%
Stabilizer
tert-Butyl hydroperoxide

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text