Ethyl N-hydroxyacetimidate, 97%
Ethyl N-hydroxyacetimidate, 97%
Ethyl N-hydroxyacetimidate, 97%
Thermo Scientific Chemicals

Ethyl N-hydroxyacetimidate, 97%

CAS: 10576-12-2 | C4H9NO2 | 103.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 197500500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10576-12-2
IUPAC Name(E)-(ethyl N-hydroxyethanimidate)
Molecular FormulaC4H9NO2
InChI KeyQWKAVVNRCKPKNM-SNAWJCMRSA-N
SMILESCCO\C(C)=N\O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid after melting
Appearance (Color)Colorless to yellow
Infrared spectrumConforms
Refractive index1.4320 to 1.4350 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only