Magnesium chloride hexahydrate, 99%, for analysis
Magnesium chloride hexahydrate, 99%, for analysis
Magnesium chloride hexahydrate, 99%, for analysis
Thermo Scientific Chemicals

Magnesium chloride hexahydrate, 99%, for analysis

CAS: 7791-18-6 | Cl2H12MgO6 | 203.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
25 kg
Packaging:
View all
Plastic drum
Catalog number 197530050
Price (EUR)
221,00
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
221,00
Each
Chemical Identifiers
CAS7791-18-6
IUPAC Namemagnesium(2+) hexahydrate dichloride
Molecular FormulaCl2H12MgO6
InChI KeyDHRRIBDTHFBPNG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Ammonium (NH4)=<50 ppm
Barium (Ba)=<20 ppm
Copper (Cu)=<10 ppm
Potassium (K)=<3000 ppm
Zinc (Zn)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only