Thermo Scientific Chemicals

Sodium hexachloroiridate(IV) hexahydrate, 99.9+%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 197570010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Sodium hexachloroiridate(IV) hexahydrate, 99.9+%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 197570010
1 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
19567-78-3
IUPAC Name
disodium hexachloroiridiumdiuide hexahydrate
Molecular Formula
Cl6H12IrNa2O6
InChI Key
QZVJPVJIAOOTJU-UHFFFAOYSA-H
SMILES
O.O.O.O.O.O.[Na+].[Na+].Cl[Ir--](Cl)(Cl)(Cl)(Cl)Cl
Iridium (Ir)
33.9 to 34.8 %
Appearance (Color)
Red to black
Purity
>=99.9 % (Metals purity) Typical
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text