Potassium hexachloroosmate(IV), min 39% Os
Potassium hexachloroosmate(IV), min 39% Os
Potassium hexachloroosmate(IV), min 39% Os
Thermo Scientific Chemicals

Potassium hexachloroosmate(IV), min 39% Os

CAS: 16871-60-6 | Cl6K2Os | 481.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 197650010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16871-60-6
IUPAC Nameosmium(4+) dipotassium hexachloride
Molecular FormulaCl6K2Os
InChI KeyVGKQJCSDERXWRV-UHFFFAOYSA-H
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[K+].[K+].[Os+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Bordeaux to purple
Appearance (Form)Powder
Assay>=39 % (Os)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only