Silver nitrate, 99.85%, for analysis
Silver nitrate, 99.85%, for analysis
Silver nitrate, 99.85%, for analysis
Thermo Scientific Chemicals

Silver nitrate, 99.85%, for analysis

CAS: 7761-88-8 | AgNO3 | 169.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 197680250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7761-88-8
IUPAC Namesilver(1+) nitrate
Molecular FormulaAgNO3
InChI KeySQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES[Ag+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Copper (Cu)=<1 ppm
Loss on drying=<0.1 % (1 g, 70°C, 2 h)
Chloride (Cl)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only