Iron, 99%, powder, -70 mesh (<212 micron)
Iron, 99%, powder, -70 mesh (&lt;212 micron)
Iron, 99%, powder, -70 mesh (&lt;212 micron)
Iron, 99%, powder, -70 mesh (&lt;212 micron)
Thermo Scientific Chemicals

Iron, 99%, powder, -70 mesh (<212 micron)

CAS: 7439-89-6 | Fe | 55.85 g/mol
数量:
500 g
2 kg
10 kg
パッケージ:
Plastic bottle
製品番号(カタログ番号) 197810020
価格(JPY)
-
見積もりを依頼する
数量:
2 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7439-89-6
IUPAC Nameiron
Molecular FormulaFe
InChI KeyXEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES[Fe]
さらに表示
Nickel (Ni)=<500 ppm
Sulfur (S)=<200 ppm
Appearance (Form)Powder
Copper (Cu)=<100 ppm
Zinc (Zn)=<50 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only