Thermo Scientific Chemicals

4-Bromobiphenyl, 99%, Thermo Scientific Chemicals

Catalog number: 197981000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Bromobiphenyl, 99%, Thermo Scientific Chemicals

Catalog number: 197981000
100 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
92-66-0
IUPAC Name
4-bromo-1,1'-biphenyl
Molecular Formula
C12H9Br
InChI Key
PKJBWOWQJHHAHG-UHFFFAOYSA-N
SMILES
BrC1=CC=C(C=C1)C1=CC=CC=C1
Infrared spectrum
Conforms
Appearance (Color)
White to light yellow
Melting point
87°C to 93°C
GC
>=98.0 %
Appearance (Form)
Crystalline powder or flakes

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text