Thermo Scientific Chemicals

Selenium, 99.5+%, powder, 200 mesh, Thermo Scientific Chemicals

Catalog number: 198070500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Selenium, 99.5+%, powder, 200 mesh, Thermo Scientific Chemicals

Catalog number: 198070500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 198070500
Price (USD)
Price: 55.90
Online price: 47.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7782-49-2
IUPAC Name
selenium
Molecular Formula
Se
InChI Key
BUGBHKTXTAQXES-UHFFFAOYSA-N
SMILES
[Se]
Appearance (Color)
Dark grey to black
Particle size
=<200 mesh
Appearance (Form)
Powder
Arsenic (As)
=<100 ppm
Iron (Fe)
=<200 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text