Hydriodic acid, for analysis, 57 wt.% aqueous solution, distilled, stabilized
Hydriodic acid, for analysis, 57 wt.%  aqueous solution, distilled, stabilized
Thermo Scientific Chemicals

Hydriodic acid, for analysis, 57 wt.% aqueous solution, distilled, stabilized

CAS: 10034-85-2 | HI | 127.91 g/mol
数量:
50 mL
250 mL
1 L
製品番号(カタログ番号) 198372500
価格(JPY)
-
見積もりを依頼する
数量:
250 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10034-85-2
IUPAC Namehydrogen iodide
Molecular FormulaHI
InChI KeyXMBWDFGMSWQBCA-UHFFFAOYSA-N
SMILESI
さらに表示
Appearance (Form)Solution
Stabilizer0.75 % H3PO2
Appearance (Color)Clear light yellow to amber
AppearanceMay darken on storage
Titration with NaOH>=55.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only