Sodium periodate, 99%, for analysis
Sodium periodate, 99%, for analysis
Sodium periodate, 99%, for analysis
Thermo Scientific Chemicals

Sodium periodate, 99%, for analysis

CAS: 7790-28-5 | INaO4 | 213.89 g/mol
数量:
500 g
2.5 kg
パッケージ:
すべて表示
Plastic Bottle
製品番号(カタログ番号) 198380025
価格(JPY)
-
数量:
2.5 kg
パッケージ:
Plastic Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7790-28-5
IUPAC Namesodium periodate
Molecular FormulaINaO4
InChI KeyJQWHASGSAFIOCM-UHFFFAOYSA-M
SMILES[Na+].[O-][I](=O)(=O)=O
さらに表示
Appearance (Color)White to off-white
Titration Iodometric>=98.5 %
pH3.4 to 4.5 (5 % at 20°C)
Sulfate (SO4)=<200 ppm
Iron (Fe)=<10 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent for analysis
RUO – Research Use Only