Sodium periodate, 99%, for analysis
Sodium periodate, 99%, for analysis
Sodium periodate, 99%, for analysis
Thermo Scientific Chemicals

Sodium periodate, 99%, for analysis

CAS: 7790-28-5 | INaO4 | 213.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 198385000
Price (USD)
436.65
Special offer
Online exclusive
Ends: 28-Dec-2026
513.00
Save 76.35
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
436.65
Special offer
Online exclusive
Ends: 28-Dec-2026
513.00
Save 76.35
Each
Chemical Identifiers
CAS7790-28-5
IUPAC Namesodium periodate
Molecular FormulaINaO4
InChI KeyJQWHASGSAFIOCM-UHFFFAOYSA-M
SMILES[Na+].[O-][I](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Titration Iodometric>=98.5 %
pH3.4 to 4.5 (5 % at 20°C)
Sulfate (SO4)=<200 ppm
Iron (Fe)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent for analysis
RUO – Research Use Only