Selenious acid, 98%
Selenious acid, 98%
Selenious acid, 98%
Thermo Scientific Chemicals

Selenious acid, 98%

CAS: 7783-00-8 | H2O3Se | 128.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 198872500
Price (USD)
229.65
Online Exclusive
255.00
Save 25.35 (10%)
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
229.65
Online Exclusive
255.00
Save 25.35 (10%)
Each
Chemical Identifiers
CAS7783-00-8
IUPAC Nameselenous acid
Molecular FormulaH2O3Se
InChI KeyMCAHWIHFGHIESP-UHFFFAOYSA-N
SMILESO[Se](O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white to light pink
Appearance (Form)Crystalline powder or crystals and/or chunks
Titration Iodometric>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only