Boron trichloride, 1M solution in hexane, AcroSeal™
Boron trichloride, 1M solution in hexane, AcroSeal™
Boron trichloride, 1M solution in hexane, AcroSeal™
Boron trichloride, 1M solution in hexane, AcroSeal™
Acros Organics

Boron trichloride, 1M solution in hexane, AcroSeal™

CAS: 10294-34-5 | BCl3 | 117.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 198921000
HSN 28121090
Price (INR)
10,611.00
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (INR)
10,611.00
Each
Chemical Identifiers
CAS10294-34-5
IUPAC Nametrichloroborane
Molecular FormulaBCl3
InChI KeyFAQYAMRNWDIXMY-UHFFFAOYSA-N
SMILESClB(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric0.9 to 1.1 M
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only