Cerium(IV) sulfate tetrahydrate, 98+%
Cerium(IV) sulfate tetrahydrate, 98+%
Cerium(IV) sulfate tetrahydrate, 98+%
Thermo Scientific Chemicals

Cerium(IV) sulfate tetrahydrate, 98+%

CAS: 10294-42-5 | CeO8S2·4H2O | 404.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 199135000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS10294-42-5
IUPAC Namecerium(3+) tetrahydrate hydrogen sulfate sulfate
Molecular FormulaCeH9O12S2
InChI KeyZBNJQJDKRZKDSV-UHFFFAOYSA-K
SMILESO.O.O.O.[Ce+3].OS([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<30 ppm
Chloride (Cl)=<100 ppm
Iron (Fe)=<150 ppm
Appearance (Color)Bright yellow
Titration Iodometric>=98.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only