Lanthanum chloride, hydrated, 64.5-70% LaCl3
Lanthanum chloride, hydrated, 64.5-70% LaCl3
Lanthanum chloride, hydrated, 64.5-70% LaCl3
Lanthanum chloride, hydrated, 64.5-70% LaCl3
Lanthanum chloride, hydrated, 64.5-70% LaCl3
Thermo Scientific Chemicals

Lanthanum chloride, hydrated, 64.5-70% LaCl3

CAS: 20211-76-1 | Cl3La | 245.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 199150025
Price (USD)
1,249.00
Each
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
1,249.00
Each
Chemical Identifiers
CAS20211-76-1
IUPAC Namelanthanum(3+) trichloride
Molecular FormulaCl3La
InChI KeyICAKDTKJOYSXGC-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[La+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Adhering crystals
Gravimetry64.5 to 70 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only