Lithium aluminum hydride, 1M solution in THF, AcroSeal™
Lithium aluminum hydride, 1M solution in THF, AcroSeal™
Lithium aluminum hydride, 1M solution in THF, AcroSeal™
Thermo Scientific Chemicals

Lithium aluminum hydride, 1M solution in THF, AcroSeal™

CAS: 16853-85-3 | AlH4Li | 37.95 g/mol
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 199491000
Price (JPY)
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16853-85-3
IUPAC Namelithium(1+) alumanuide
Molecular FormulaAlH4Li
InChI KeyOCZDCIYGECBNKL-UHFFFAOYSA-N
SMILES[Li+].[AlH4-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Turbid grey
Appearance (Form)Solution
Strength0.90 to 1.10 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only