Molybdate 3 Reagent Solution
Product Image
Hach Pacific Pty Ltd

Molybdate 3 Reagent Solution

For low range silica determination by the Heteropoly Blue method.
Have Questions?
Catalog NumberQuantity
19952650 mL
Catalog number 199526
Price (USD)
-
Quantity:
50 mL
For low range silica determination by the Heteropoly Blue method. 50 mL Self-Contained Dropping Bottle.
    Specifications

  • Method Name: Heteropoly Blue
  • Number of tests: 50
  • Parameter: Silica
  • Platform: Solution
  • Quantity: 50 mL
Specifications
Quantity50 mL
TypeSolution
Volume (Metric)50 mL
Unit SizeEach